4-Brombenzolsulfonamid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Brombenzolsulfonsäureamid

OR30462-25G OR30462-5G OR30462-100G
APOSOR30462-25GEA 26.1 EUR
APOSOR30462-25G APOSOR30462-5G APOSOR30462-100G
4-Brombenzolsulfonamid 98%
4-Brombenzolsulfonamid
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,09 g/mol
Schmelzpunkt: 164…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051977
CAS-Nummer: 701-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR