2,5-Dibromhydrochinon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,5-Dibromobenzene-1,4-diol
OR30490-10G
OR30490-5G
APOSOR30490-10GEA
33.5
EUR
APOSOR30490-10G
APOSOR30490-5G
2,5-Dibromhydrochinon 98%
2,5-Dibromhydrochinon
Formel:
C₆H₄Br₂O₂ Schmelzpunkt: 193 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192664 CAS-Nummer: 14753-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...