7-Isochinolinol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Hydroxyisoquinoline, Isoquinolin-7-ol
OR30506-5G
OR30506-25G
APOSOR30506-5GEA
172
EUR
APOSOR30506-5G
APOSOR30506-25G
7-Isochinolinol 98%
7-Isochinolinol
Formel:
C₉H₇NO Molecular Weight: 145.16 g/mol Schmelzpunkt: 226…228 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00456131 CAS-Nummer: 7651-83-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...