3-Iod-1H-indazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Iodoindazole
OR305076-1G
OR305076-5G
APOSOR305076-1GEA
21.9
EUR
APOSOR305076-1G
APOSOR305076-5G
3-Iod-1H-indazol 98%
3-Iodo-1H-indazole
Formel:
C₇H₅IN₂ Molecular Weight: 244,0325 g/mol Schmelzpunkt: 139…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972469 CAS-Nummer: 66607-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...