2-Brom-m-toluylsäure

Lieferant: Apollo Scientific
Achtung

OR3051-25G OR3051-100G
APOSOR3051-25GEA 26.1 EUR
APOSOR3051-25G APOSOR3051-100G
2-Brom-m-toluylsäure
2-Brom-m-toluylsäure
Formel: C₈H₇BrO₂
Molecular Weight: 215.05 g/mol
Schmelzpunkt: 134…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079721
CAS-Nummer: 53663-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR