5-Bromo-1H-indazol-3-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-1,2-dihydro-3H-indazol-3-one

OR305112-1G
APOSOR305112-1GEA 84.1 EUR
APOSOR305112-1G
5-Bromo-1H-indazol-3-ol
5-Bromo-1H-indazol-3-ol
Formel: C₇H₅BrN₂O
Molecular Weight: 213,0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781638
CAS-Nummer: 7364-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR