3-Iodimidazo[1,2-a]pyridin 95+%
Lieferant: Apollo Scientific
OR305157-5G
APOSOR305157-5GEA
121
EUR
APOSOR305157-5G
3-Iodimidazo[1,2-a]pyridin 95+%
3-Iodimidazo[1,2-a]pyridin
Formel:
C₇H₅IN₂ Molecular Weight: 244,0300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06739222 CAS-Nummer: 307503-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...