2-Chloro-1H-indole-3-carboxylic acid

Lieferant: Apollo Scientific

OR305201-1G OR305201-500MG
APOSOR305201-1GEA 366 EUR
APOSOR305201-1G APOSOR305201-500MG
2-Chloro-1H-indole-3-carboxylic acid
2-Chloro-1H-indole-3-carboxylic acid
Formel: C₉H₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01719046
CAS-Nummer: 54778-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR