6-Bromoisoindolin-1-one 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-2,3-dihydro-1H-isoindol-1-one

OR305290-1G OR305290-5G OR305290-500MG
APOSOR305290-1GEA 37.7 EUR
APOSOR305290-1G APOSOR305290-5G APOSOR305290-500MG
6-Bromoisoindolin-1-one 97%
6-Bromoisoindolin-1-one
Formel: C₈H₆BrNO
Molecular Weight: 212.0433 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11848599
CAS-Nummer: 675109-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR