Sodium-2-naphthalenesulphonate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Naphthalenesulphonic acid Sodium Salt, Sodium 2-naphthalenesulphonate

OR30530-1KG OR30530-250G OR30530-5KG
APOSOR30530-1KGEA 65.4 EUR
APOSOR30530-1KG APOSOR30530-250G APOSOR30530-5KG
Sodium-2-naphthalenesulphonate 95%
Sodium-2-naphthalenesulphonate
Formel: C₁₀H₈O₃S
Molecular Weight: 230.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064186
CAS-Nummer: 532-02-5
EINECS: 208-523-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR