5-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 97%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-1H-pyrrolo[2,3-b]pyridin-2-one

OR305356-5G
APOSOR305356-5GEA 81.2 EUR
APOSOR305356-5G
5-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 97%
5-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
Formel: C₇H₅BrN₂O
Molecular Weight: 213,0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07367221
CAS-Nummer: 183208-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR