2-Hydrazino-3-methylpyridin

Lieferant: Apollo Scientific

Synonyme: 2-Hydrazinyl-3-methylpyridine

OR305404-1G OR305404-5G
APOSOR305404-1GEA 0 EUR
APOSOR305404-1G APOSOR305404-5G
2-Hydrazino-3-methylpyridin
2-Hydrazino-3-methylpyridin
Formel: C₆H₉N₃
Molecular Weight: 123,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07786312
CAS-Nummer: 4930-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR