1-Methyl-1H-indazol-6-boronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-1H-indazole-6-boronic Acid, 1-Methylindazole-6-boronic acid

OR30558-1G OR30558-250MG
APOSOR30558-1GEA 72.5 EUR
APOSOR30558-1G APOSOR30558-250MG
1-Methyl-1H-indazol-6-boronsäure
1-Methyl-1H-indazole-6-boronic acid
Formel: C₈H₉BN₂O₂
Molecular Weight: 175,9803 g/mol
Schmelzpunkt: 277…283 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09870053
CAS-Nummer: 1150114-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR