Isochinolin-5-carbaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Isoquinoline-5-carboxaldehyde
OR30559-1G
OR30559-250MG
APOSOR30559-1GEA
48
EUR
APOSOR30559-1G
APOSOR30559-250MG
Isochinolin-5-carbaldehyd
Isochinolin-5-carbaldehyd
Formel:
C₁₀H₇NO Molecular Weight: 157.17 g/mol Schmelzpunkt: 116…120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03412483 CAS-Nummer: 80278-67-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...