Acetamidoxime

Lieferant: Apollo Scientific
Achtung

Synonyme: Acetamide oxime

OR30585-1G OR30585-5G
APOSOR30585-1GEA 21.9 EUR
APOSOR30585-1G APOSOR30585-5G
Acetamidoxime
Acetamidoxime
Formel: C₂H₆N₂O
Molecular Weight: 74,08 g/mol
Schmelzpunkt: 134…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00603514
CAS-Nummer: 22059-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR