1-Methyl-1H-indazol-5-boronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Methyl-1H-indazol-5-yl-5-boronic acid, (1-methyl-1H-indazol-5-yl)boronic acid
OR30587-1G
OR30587-250MG
APOSOR30587-1GEA
26.1
EUR
APOSOR30587-1G
APOSOR30587-250MG
1-Methyl-1H-indazol-5-boronsäure
1-Methyl-1H-indazole-5-boronic acid
Formel:
C₈H₉BN₂O₂ Molecular Weight: 175,9800 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09870052 CAS-Nummer: 590418-08-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...