1-Methyl-1H-indazol-7-boronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1-Methyl-1H-indazol-7-yl)boronic acid
OR30588-1G
OR30588-250MG
APOSOR30588-1GEA
134
EUR
APOSOR30588-1G
APOSOR30588-250MG
1-Methyl-1H-indazol-7-boronsäure
1-Methyl-1H-indazole-7-boronic acid
Formel:
C₈H₉BN₂O₂ Molecular Weight: 175,9803 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09870054 CAS-Nummer: 1001907-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...