2-Methyl-2H-indazol-4-boronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

OR30589-250MG OR30589-1G OR30589-5G
APOSOR30589-250MGEA 60.9 EUR
APOSOR30589-250MG APOSOR30589-1G APOSOR30589-5G
2-Methyl-2H-indazol-4-boronsäurepinakolester
2-Methyl-2H-indazol-4-boronsäurepinakolester
Formel: C₁₄H₁₉BN₂O₂
Molecular Weight: 258.1239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109437
CAS-Nummer: 885698-95-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR