4-Pyridinthiol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Pyridin-4-thiol, 4-Mercaptopyridin
OR30596-1G
OR30596-5G
APOSOR30596-1GEA
21.9
EUR
APOSOR30596-1G
APOSOR30596-5G
4-Pyridinthiol 97%
4-Pyridinthiol
Formel:
C₅H₅NS Molecular Weight: 111,17 g/mol Schmelzpunkt: 182…189 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006422 CAS-Nummer: 4556-23-4 EINECS: 224-926-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...