8-Bromo-1,6-naphthyridine
Lieferant: Apollo Scientific
OR30599-1G
OR30599-50MG
OR30599-250MG
OR30599-500MG
OR30599-2.5G
OR30599-5G
APOSOR30599-1GEA
458
EUR
APOSOR30599-1G
APOSOR30599-50MG
APOSOR30599-250MG
APOSOR30599-500MG
APOSOR30599-2.5G
APOSOR30599-5G
8-Bromo-1,6-naphthyridine
8-Bromo-1,6-naphthyridine
Formel:
C₈H₅BrN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234384 CAS-Nummer: 17965-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...