5-Bromimidazo[1,2-a]pyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR30602-1G
OR30602-5G
APOSOR30602-1GEA
24.8
EUR
APOSOR30602-1G
APOSOR30602-5G
5-Bromimidazo[1,2-a]pyridin
5-Bromimidazo[1,2-a]pyridin
Formel:
C₇H₅BrN₂ Molecular Weight: 197,03 g/mol Schmelzpunkt: 61…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08275698 CAS-Nummer: 69214-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...