1H-Imidazol-2-carbonsäure 96%

Lieferant: Apollo Scientific
Achtung

OR30610-5G OR30610-25G
APOSOR30610-5GEA 109 EUR
APOSOR30610-5G APOSOR30610-25G
1H-Imidazol-2-carbonsäure 96%
2-Imidazolecarboxylic acid
Formel: C₄H₄N₂O₂
Molecular Weight: 112.09 g/mol
Schmelzpunkt: 198 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00270816
CAS-Nummer: 16042-25-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR