3-Hydrazinylpyridine dihydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR306302-5G
APOSOR306302-5GEA 84.1 EUR
APOSOR306302-5G
3-Hydrazinylpyridine dihydrochloride 95%
3-Hydrazinylpyridine dihydrochloride
Formel: C5H9Cl2N3
Molecular Weight: 182,0511 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08576202
CAS-Nummer: 364727-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR