2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid 95+%

Lieferant: Apollo Scientific

OR306578-10G OR306578-25G
APOSOR306578-10GEA 65.4 EUR
APOSOR306578-10G APOSOR306578-25G
2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid 95+%
2-Methylpyrazolo[1,5-a]pyrimidine-6-carboxylic acid
Formel: C₈H₇N₃O₂
Molecular Weight: 177.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19443280
CAS-Nummer: 739364-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR