4-Brom-2-nitroanisol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1-methoxy-2-nitrobenzene

OR3066-25G OR3066-5G OR3066-100G
APOSOR3066-25GEA 24.8 EUR
APOSOR3066-25G APOSOR3066-5G APOSOR3066-100G
4-Brom-2-nitroanisol
4-Brom-2-nitroanisol
Formel: C₇H₆BrNO₃
Molecular Weight: 232.03 g/mol
Schmelzpunkt: 87…88 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00055529
CAS-Nummer: 33696-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR