2-Cyclopropylisonicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR306659-1G
OR306659-250MG
APOSOR306659-1GEA
429
EUR
APOSOR306659-1G
APOSOR306659-250MG
2-Cyclopropylisonicotinic acid
2-Cyclopropylisonicotinic acid
Formel:
C₉H₉NO₂ Schmelzpunkt: 285…287 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13187883 CAS-Nummer: 1216171-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...