2-Chlorothiophene-3-carboxylic acid

Lieferant: Apollo Scientific

OR307013-5G
APOSOR307013-5GEA 139 EUR
APOSOR307013-5G
2-Chlorothiophene-3-carboxylic acid
2-Chlorothiophene-3-carboxylic acid
Formel: C₅H₃ClO₂S
Molecular Weight: 162,5941 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00098423
CAS-Nummer: 53935-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR