Boc-D-α-methylserine

Lieferant: Apollo Scientific
Achtung

OR307056-1G
APOSOR307056-1GEA 633 EUR
APOSOR307056-1G
Boc-D-α-methylserine
Boc-D-alpha-methylserine
Formel: C₉H₁₇NO₅
Molecular Weight: 219,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02682596
CAS-Nummer: 84311-18-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR