(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Amino-N-Boc-L-alanine, Nalpha-Boc-2,3-diaminopropionic Acid, Boc-Dpr-OH, Boc-Dap-OH, 3-Amino-N-(tert-butoxycarbonyl)-L-alanine
OR307060-5G
APOSOR307060-5GEA
36.4
EUR
APOSOR307060-5G
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid 96%
(S)-3-Amino-2-(tert-butoxycarbonylamino)propionic acid
Formel:
C₈H₁₆N₂O₄ Molecular Weight: 204,23 g/mol Schmelzpunkt: 210 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236843 CAS-Nummer: 73259-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...