Boc-trans-4-hydroxy-L-proline 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, Boc-Hyp-OH, trans-N-Boc-4-hydroxy-L-proline

OR307069-25G OR307069-100G
APOSOR307069-25GEA 24.8 EUR
APOSOR307069-25G APOSOR307069-100G
Boc-trans-4-hydroxy-L-proline 97%
Boc-trans-4-hydroxy-L-proline
Formel: C₁₀H₁₇NO₅
Molecular Weight: 231,3 g/mol
Schmelzpunkt: 123…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053370
CAS-Nummer: 13726-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR