Boc-D-Pro-OH 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid, N-Boc-D-proline
OR307074-500G
APOSOR307074-500GEA
165
EUR
APOSOR307074-500G
Boc-D-Pro-OH 97%
Boc-D-Pro-OH
Formel:
C₁₀H₁₇NO₄ Molecular Weight: 215,25 g/mol Schmelzpunkt: 134…137 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00063226 CAS-Nummer: 37784-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...