Boc-O-methyl-L-serine

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-2-amino-3-methoxypropionic acid, Boc-Ser(Me)-OH, N-tert-Butoxycarbonyl-O-methyl-L-serine, (2S)-2-Amino-3-methoxypropanoic acid, N-BOC protected

OR307095-1G OR307095-5G
APOSOR307095-1GEA 21.9 EUR
APOSOR307095-1G APOSOR307095-5G
Boc-O-methyl-L-serine
Boc-O-methyl-L-serine
Formel: C₉H₁₇NO₅
Molecular Weight: 219.237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153314
CAS-Nummer: 51293-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR