1-Methyl-1H-indazole-4-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-1H-indazole-4-carbaldehyde

OR30711-500MG OR30711-1G
APOSOR30711-500MGEA 306 EUR
APOSOR30711-500MG APOSOR30711-1G
1-Methyl-1H-indazole-4-carboxaldehyde
1-Methyl-1H-indazole-4-carboxaldehyde
Formel: C₉H₈N₂O
Molecular Weight: 160,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10699167
CAS-Nummer: 1053655-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR