Boc-Abu-OH 97+%

Lieferant: Apollo Scientific

Synonyme: (S)-2-(tert-Butoxycarbonylamino)butyric acid, Boc-L-2-aminobutanoic acid, BOC-L-α-Aminobutyric acid, Boc-L-Abu-OH, (S)-2-(Boc-amino)butyric acid

OR307113-500G
APOSOR307113-500GEA 119 EUR
APOSOR307113-500G
Boc-Abu-OH 97+%
Boc-Abu-OH
Formel: C₉H₁₇NO₄
Molecular Weight: 203,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037267
CAS-Nummer: 34306-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR