Methyl-1H-indazol-4-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1H-indazole-4-carboxylate

OR30714-1G OR30714-5G OR30714-25G
APOSOR30714-1GEA 21.9 EUR
APOSOR30714-1G APOSOR30714-5G APOSOR30714-25G
Methyl-1H-indazol-4-carboxylat
Methyl-1H-indazol-4-carboxylat
Formel: C₉H₈N₂O₂
Molecular Weight: 176,1700 g/mol
Schmelzpunkt: 133…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07371610
CAS-Nummer: 192945-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR