Methyl-1H-indazol-7-carboxylat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1H-indazole-7-carboxylate

OR30717-5G OR30717-25G
APOSOR30717-5GEA 60.9 EUR
APOSOR30717-5G APOSOR30717-25G
Methyl-1H-indazol-7-carboxylat 98%
Methyl-1H-indazol-7-carboxylat
Formel: C₉H₈N₂O₂
Molecular Weight: 176,1720 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371608
CAS-Nummer: 755752-82-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR