1-Ethyl-3-nitro-1H-pyrazole

Lieferant: Apollo Scientific

OR307177-1G OR307177-250MG OR307177-500MG
APOSOR307177-1GEA 510 EUR
APOSOR307177-1G APOSOR307177-250MG APOSOR307177-500MG
1-Ethyl-3-nitro-1H-pyrazole
1-Ethyl-3-nitro-1H-pyrazole
Formel: C₅H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04970018
CAS-Nummer: 58793-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR