Methyl-1-methyl-1H-indazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1-methyl-1H-indazole-4-carboxylate

OR30718-1G OR30718-250MG
APOSOR30718-1GEA 33.5 EUR
APOSOR30718-1G APOSOR30718-250MG
Methyl-1-methyl-1H-indazole-4-carboxylate
Methyl-1-methyl-1H-indazole-4-carboxylate
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190.1986 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11109402
CAS-Nummer: 1071428-42-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR