5-Phenyl-1H-[1,2,4]triazol-3-ylamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Phenyl-1H-1,2,4-triazol-3-amine, 3-Phenyl-1H-1,2,4-triazol-5-amine
OR307244-1G
OR307244-250MG
APOSOR307244-1GEA
128
EUR
APOSOR307244-1G
APOSOR307244-250MG
5-Phenyl-1H-[1,2,4]triazol-3-ylamine
5-Phenyl-1H-[1,2,4]triazol-3-ylamine
Formel:
C₈H₈N₄ Molecular Weight: 160.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00086332 CAS-Nummer: 4922-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...