4-Iodo-3-nitro-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR307289-1G OR307289-5G
APOSOR307289-1GEA 145 EUR
APOSOR307289-1G APOSOR307289-5G
4-Iodo-3-nitro-1H-pyrazole
4-Iodo-3-nitro-1H-pyrazole
Formel: C₃H₂IN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01821344
CAS-Nummer: 400753-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR