4-Chloro-3-nitro-1H-pyrazole 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR307294-1G
OR307294-5G
APOSOR307294-1GEA
125
EUR
APOSOR307294-1G
APOSOR307294-5G
4-Chloro-3-nitro-1H-pyrazole 95%
4-Chloro-3-nitro-1H-pyrazole
Formel:
C₃H₂ClN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01114991 CAS-Nummer: 400752-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...