1H-Indazole-5-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR30733-1G OR30733-250MG
APOSOR30733-1GEA 72.5 EUR
APOSOR30733-1G APOSOR30733-250MG
1H-Indazole-5-carbonitrile
1H-Indazole-5-carbonitrile
Formel: C₈H₅N₃
Molecular Weight: 143,1454 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04973397
CAS-Nummer: 74626-47-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR