(3-Bromphenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Bromophenyl)acetic acid

OR3074-100G OR3074-25G
APOSOR3074-100GEA 68.3 EUR
APOSOR3074-100G APOSOR3074-25G
(3-Bromphenyl)essigsäure 98%
(3-Bromphenyl)essigsäure
Formel: (C₆H₄Br)CH₂COOH
Molecular Weight: 215,05 g/mol
Schmelzpunkt: 98…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004330
CAS-Nummer: 1878-67-7
EINECS: 217-522-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR