4-Brom-8-chlorchinolin

Lieferant: Apollo Scientific

OR307510-1G OR307510-5G
APOSOR307510-1GEA 243 EUR
APOSOR307510-1G APOSOR307510-5G
4-Brom-8-chlorchinolin
4-Bromo-8-chloroquinoline
Formel: C₉H₅BrClN
Lagertemperatur: Raumtemperatur
MDL: MFCD07700233
CAS-Nummer: 927800-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR