2-Amino-6-methoxy-3-methylchinolin

Lieferant: Apollo Scientific
Achtung

OR307521-1G OR307521-5G
APOSOR307521-1GEA 343 EUR
APOSOR307521-1G APOSOR307521-5G
2-Amino-6-methoxy-3-methylchinolin
2-Amino-6-methoxy-3-methylquinoline
Formel: C₁₁H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00272514
CAS-Nummer: 203506-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR