2-Amino-4-methylchinolin Hydrochlorid
Lieferant: Apollo Scientific
OR307522-1G
OR307522-5G
APOSOR307522-1GEA
335
EUR
APOSOR307522-1G
APOSOR307522-5G
2-Amino-4-methylchinolin Hydrochlorid
2-Amino-4-methylchinolin Hydrochlorid
Formel:
C₁₀H₁₁ClN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07700220 CAS-Nummer: 1171502-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...