4-Amino-6,8-dimethylchinolin

Lieferant: Apollo Scientific

OR307525-1G OR307525-5G
APOSOR307525-1GEA 219 EUR
APOSOR307525-1G APOSOR307525-5G
4-Amino-6,8-dimethylchinolin
4-Amino-6,8-dimethylquinoline
Formel: C₁₁H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08754057
CAS-Nummer: 929339-38-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR