4-Bromo-1-methylindolin-2-one
Lieferant: Apollo Scientific
|
Achtung
|
OR30753-1G
OR30753-250MG
APOSOR30753-1GEA
609
EUR
APOSOR30753-1G
APOSOR30753-250MG
4-Bromo-1-methylindolin-2-one
4-Bromo-1-methylindolin-2-one
Formel:
C₉H₈BrNO Molecular Weight: 226,08 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD10699229 CAS-Nummer: 884855-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...