Boc-3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid
Lieferant: Apollo Scientific
Synonyme:
diexo-3-tert-Butoxycarbonylamino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
OR307547-1G
OR307547-250MG
APOSOR307547-1GEA
872
EUR
APOSOR307547-1G
APOSOR307547-250MG
Boc-3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid
Boc-3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid
Formel:
C₁₃H₁₉NO₄ Molecular Weight: 253.3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682617 CAS-Nummer: 148257-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...