diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide

Lieferant: Apollo Scientific

Synonyme: 3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxamide

OR307551-1G OR307551-250MG
APOSOR307551-1GEA 665 EUR
APOSOR307551-1G APOSOR307551-250MG
diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide
diexo-3-Amino-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid amide
Formel: C₈H₁₂N₂O
Molecular Weight: 152.197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00143157
CAS-Nummer: 105786-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR